Product Details
- SNP ID
-
rs3844454
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.19:53911001 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- TGAATCCTAGTTGGAAGGATGAGAG[A/T]CCGTTGTTTGACTGGGGTGCTCTGA
- Phenotype
-
MIM: 606899
MIM: 176980
- Polymorphism
- A/T, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
CACNG7
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs148889389,rs78085696] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- CACNG7
- Gene Name
- calcium voltage-gated channel auxiliary subunit gamma 7
- Gene
- PRKCG
- Gene Name
- protein kinase C gamma
There are no transcripts associated with this gene.
View Full Product Details