Product Details

SNP ID
rs768659
Assay Type
Validated
NCBI dbSNP Submissions
55
Location
Chr.1:37697035 on Build GRCh38
Set Membership
HapMap Validated
Context Sequence [VIC/FAM]
TGCAGTTCTCTCTTTTTAAATTAGA[A/C]ATACAGAACTGCAAATGAGAACTTC
Phenotype
MIM: 614799 MIM: 609977
Polymorphism
A/C, Transversion Substitution
Allele Nomenclature
Literature Links
C1orf109 PubMed Links
Additional Information
For this assay, SNP(s) [rs57424477] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.

Gene Details

Gene
C1orf109
Gene Name
chromosome 1 open reading frame 109
There are no transcripts associated with this gene.

Gene
CDCA8
Gene Name
cell division cycle associated 8
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001256875.1 Intron NP_001243804.1
NM_018101.3 Intron NP_060571.1

View Full Product Details