Product Details
- SNP ID
-
rs375994
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.5:7402296 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- AGTTAGCTCATGGGTTTGGTCCATT[G/T]GACAAAATTAAGCCAGTGCTAATTC
- Phenotype
-
MIM: 103071
- Polymorphism
- G/T, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
ADCY2
PubMed Links
Gene Details
- Gene
- ADCY2
- Gene Name
- adenylate cyclase 2
View Full Product Details