Product Details

SNP ID
rs592127
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.10:95201340 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TCCCCTCCAGCCTCCCCAATGTCTA[C/T]TATTTCCATCTTTATGTCCATATGT
Phenotype
Polymorphism
C/T, Transition Substitution
Allele Nomenclature
Literature Links
ACSM6 PubMed Links
Additional Information
For this assay, SNP(s) [rs141500188] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.

Gene Details

Gene
ACSM6
Gene Name
acyl-CoA synthetase medium-chain family member 6
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_207321.2 Intron NP_997204.2

View Full Product Details