Product Details
- SNP ID
-
rs296364
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.19:47872441 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- TCTCCGTCACTCTACTCACACGGGG[A/G]CTTCTGATTCTGCCTCAAACGTGGC
- Phenotype
-
MIM: 125263
- Polymorphism
- A/G, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
LINC01595
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs62529871] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- LINC01595
- Gene Name
- long intergenic non-protein coding RNA 1595
There are no transcripts associated with this gene.
- Gene
- SULT2A1
- Gene Name
- sulfotransferase family 2A member 1
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_003167.3 |
|
Intron |
|
|
NP_003158.2 |
View Full Product Details