Product Details

SNP ID
rs135624
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.22:44494805 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CAAAAAGCCACTCGAGTAGAGTGAG[C/G]GCTGGCGACACAGCAGAGTTCCCAC
Phenotype
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
LDOC1L PubMed Links

Gene Details

Gene
LDOC1L
Gene Name
leucine zipper down-regulated in cancer 1 like
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_032287.2 3089 UTR 3 NP_115663.2

View Full Product Details