Product Details

SNP ID
rs34783504
Assay Type
Functionally tested
NCBI dbSNP Submissions
13
Location
Chr.1:23633561 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCGTGATCAAAGAATATGCCTTCCT[A/G]GGAGAATAAGCGGGTAGTGACAGCC
Phenotype
MIM: 607305
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
MDS2 PubMed Links

Gene Details

Gene
MDS2
Gene Name
myelodysplastic syndrome 2 translocation associated
There are no transcripts associated with this gene.

View Full Product Details