Product Details

SNP ID
rs12670393
Assay Type
Functionally Tested
NCBI dbSNP Submissions
31
Location
Chr.7:2633213 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCCGTGACCAGGGTCCCTGGTCACC[C/T]GGAGTTTCTCTCTGGACGGATCATG
Phenotype
MIM: 608919
Polymorphism
C/T, Transition Substitution
Allele Nomenclature
Literature Links
TTYH3 PubMed Links

Gene Details

Gene
TTYH3
Gene Name
tweety family member 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_025250.2 Intron NP_079526.1
XM_011515535.2 Intron XP_011513837.1
XM_017012655.1 Intron XP_016868144.1
XM_017012656.1 Intron XP_016868145.1

View Full Product Details