Product Details
- SNP ID
-
rs12357316
- Assay Type
- Validated
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.10:74000944 on Build GRCh38
- Set Membership
-
Validated
- Context Sequence [VIC/FAM]
- AGTAGGGAGGATTGTGCTGATAAAT[A/G]GTTAATCATGAACAAAGGTACTTGT
- Phenotype
-
MIM: 193065
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
VCL
PubMed Links
Gene Details
- Gene
- VCL
- Gene Name
- vinculin
View Full Product Details