Product Details

SNP ID
rs9848929
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:167442535 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATAGCATAATAAAACGGTGATGAGC[A/G]TGGTAGAGTTTTAAATTTAAGGCAT
Phenotype
MIM: 605587
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
SERPINI2 PubMed Links

Gene Details

Gene
SERPINI2
Gene Name
serpin family I member 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001012303.2 Intron NP_001012303.1
NM_006217.4 Intron NP_006208.1

View Full Product Details