Product Details

SNP ID
rs12542104
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:10606797 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CCTTGGTCTTTGTCCATGTACTATG[A/G]ACATCTCCAGTGGACTGAACGTTGC
Phenotype
MIM: 608581
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
RP1L1 PubMed Links

Gene Details

Gene
RP1L1
Gene Name
retinitis pigmentosa 1-like 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_178857.5 7530 UTR 3 NP_849188.4

View Full Product Details