Product Details

SNP ID
rs34416776
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:138496935 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCCTATTAAAATATGTTTATTAATG[G/T]GCCAAATATTTATGAATGATGAGAA
Phenotype
MIM: 614310
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
CEP70 PubMed Links

Gene Details

Gene
CEP70
Gene Name
centrosomal protein 70
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001288964.1 2139 Intron NP_001275893.1
NM_001288965.1 2139 Intron NP_001275894.1
NM_001288966.1 2139 Intron NP_001275895.1
NM_001288967.1 2139 Intron NP_001275896.1
NM_001320598.1 2139 Intron NP_001307527.1
NM_001320599.1 2139 UTR 3 NP_001307528.1
NM_001320600.1 2139 Intron NP_001307529.1
NM_024491.3 2139 Intron NP_077817.2
XM_005247805.3 2139 UTR 3 XP_005247862.1
XM_011513209.2 2139 Intron XP_011511511.1
XM_017007270.1 2139 Intron XP_016862759.1
XM_017007271.1 2139 Intron XP_016862760.1
XM_017007272.1 2139 Intron XP_016862761.1
XM_017007273.1 2139 Intron XP_016862762.1
XM_017007274.1 2139 Intron XP_016862763.1
XM_017007275.1 2139 Intron XP_016862764.1
XM_017007276.1 2139 Intron XP_016862765.1
XM_017007277.1 2139 Intron XP_016862766.1
XM_017007278.1 2139 Intron XP_016862767.1
XM_017007279.1 2139 Intron XP_016862768.1
XM_017007280.1 2139 Intron XP_016862769.1
XM_017007281.1 2139 Intron XP_016862770.1
XM_017007282.1 2139 Intron XP_016862771.1
XM_017007283.1 2139 Intron XP_016862772.1
XM_017007284.1 2139 Intron XP_016862773.1

View Full Product Details