Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_001199797.1 | 2133 | UTR 3 | NP_001186726.1 | ||
NM_002832.3 | 2133 | UTR 3 | NP_002823.3 | ||
NM_080588.2 | 2133 | UTR 3 | NP_542155.1 | ||
XM_011509819.1 | 2133 | UTR 3 | XP_011508121.1 | ||
XM_011509820.2 | 2133 | UTR 3 | XP_011508122.1 | ||
XM_011509821.1 | 2133 | UTR 3 | XP_011508123.1 | ||
XM_011509822.2 | 2133 | UTR 3 | XP_011508124.1 | ||
XM_011509823.1 | 2133 | Intron | XP_011508125.1 | ||
XM_011509824.1 | 2133 | UTR 3 | XP_011508126.1 | ||
XM_017001935.1 | 2133 | UTR 3 | XP_016857424.1 | ||
XM_017001936.1 | 2133 | UTR 3 | XP_016857425.1 | ||
XM_017001937.1 | 2133 | UTR 3 | XP_016857426.1 |