Product Details

SNP ID
rs12138972
Assay Type
Functionally tested
NCBI dbSNP Submissions
43
Location
Chr.1:182904296 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TCTAGAGTCATGTGACCAGACTCCA[C/T]CACCACGATACCATCAACCGTCCCT
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
SHCBP1L PubMed Links

Gene Details

Gene
SHCBP1L
Gene Name
SHC binding and spindle associated 1 like
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_030933.2 1756 Missense Mutation ATG,GTG M491V NP_112195.2

View Full Product Details