Product Details

SNP ID
rs7076948
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:46037697 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TCTGCCTATGCTCAGAGCCACTGCT[G/A]TTTCTACAAATCTACAAGGCCCAGA
Phenotype
MIM: 157145 MIM: 601984
Polymorphism
G/A, Transition substitution
Allele Nomenclature
Literature Links
MSMB PubMed Links

Gene Details

Gene
MSMB
Gene Name
microseminoprotein beta
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002443.3 Intron NP_002434.1
NM_138634.2 Intron NP_619540.1
Gene
NCOA4
Gene Name
nuclear receptor coactivator 4
There are no transcripts associated with this gene.

View Full Product Details