Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
sprr-2
Entrez Gene ID
181575
Gene Name
Sex peptide receptor-related protein 2
Gene Aliases
CELE_F42D1.3
Chromosome Location
Chr.X: 14793445 - 14804359 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_078054.7 NP_510455.2 3 - 4 277 81

View Full Product Details