Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
sop-3
Entrez Gene ID
171840
Gene Name
protein-coding
Gene Aliases
CELE_Y71F9B.10
Chromosome Location
Chr.I: 2761158 - 2776104 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001306444.3 NP_001293373.1 13 - 14 4269 92
NM_001306445.3 NP_001293374.1 8 - 9 3284 92
NM_001306446.3 NP_001293375.1 8 - 9 3302 92
NM_001306447.3 NP_001293376.1 1 - 2 227 92
NM_001377813.3 NP_001364767.1 14 - 15 3956 92
NM_001377816.2 NP_001364504.1 14 - 15 3978 92
NM_170837.5 NP_740821.1 13 - 14 4254 92
GenBank mRNA AF308860.1 - 13 - 14 4253 92

View Full Product Details