Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
sop-3
Entrez Gene ID
171840
Gene Name
protein-coding
Gene Aliases
CELE_Y71F9B.10
Chromosome Location
Chr.I: 2761158 - 2776104 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001306444.3 NP_001293373.1 2 - 3 290 126
NM_001377813.3 NP_001364767.1 2 - 3 289 126
NM_001377816.2 NP_001364504.1 2 - 3 293 126
NM_170837.5 NP_740821.1 2 - 3 293 126
GenBank mRNA AF308860.1 - 2 - 3 292 126

View Full Product Details