Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
hrpf-1
Entrez Gene ID
172395
Gene Name
protein-coding
Gene Aliases
CELE_W02D3.11
Chromosome Location
Chr.I: 6753219 - 6756354 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001372997.1 NP_001359979.1 4 - 5 1047 84

View Full Product Details