Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
apg-1
Entrez Gene ID
173304
Gene Name
AP-1 complex subunit gamma
Gene Aliases
CELE_Y105E8A.9
Chromosome Location
Chr.I: 14406028 - 14417942 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_170941.7 NP_740937.1 3 - 4 414 69

View Full Product Details