Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
dhs-6
Entrez Gene ID
3565470
Gene Name
SCP2 domain-containing protein
Gene Aliases
CELE_C17G10.8
Chromosome Location
Chr.II: 5617122 - 5619319 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001026801.6 NP_001021972.1 5 - 6 1045 125

View Full Product Details