Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
pef-1
Entrez Gene ID
175257
Gene Name
protein-coding
Gene Aliases
CELE_F23H11.8
Chromosome Location
Chr.III: 919274 - 926316 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001381720.1 NP_001367778.1 2 - 3 312 137
NM_171080.6 NP_741091.1 2 - 3 310 137
GenBank mRNA AF023454.1 - 2 - 3 338 137

View Full Product Details