Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
frm-8
Entrez Gene ID
175428
Gene Name
protein-coding
Gene Aliases
CELE_H09G03.2
Chromosome Location
Chr.III: 3200795 - 3219150 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_065271.6 NP_497672.2 6 - 7 1096 66
NM_171096.6 NP_741109.1 7 - 8 1220 66
NM_171097.5 NP_741110.1 6 - 7 1108 66

View Full Product Details