Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
inx-18
Entrez Gene ID
182801
Gene Name
protein-coding
Gene Aliases
CELE_C18H7.2
Chromosome Location
Chr.IV: 606355 - 614062 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_171246.5 NP_741294.1 2 - 3 252 74
NM_171247.6 NP_741295.1 2 - 3 234 74

View Full Product Details