Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ttr-55
Entrez Gene ID
186397
Gene Name
TransThyretin-Related family domain
Gene Aliases
CELE_F56F4.2
Chromosome Location
Chr.I: 6145455 - 6146477 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_059371.5 NP_491772.4 2 - 3 212 86

View Full Product Details