Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
cof-2
Entrez Gene ID
183582
Gene Name
Olfactomedin-like domain-containing protein
Gene Aliases
CELE_C48E7.5
Chromosome Location
Chr.I: 6250475 - 6252876 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_059403.5 NP_491804.2 9 - 10 1349 108
GenBank mRNA AY494975.1 - 9 - 10 1253 108

View Full Product Details