Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
snt-4
Entrez Gene ID
172346
Gene Name
SyNapTotagmin
Gene Aliases
CELE_T23H2.2
Chromosome Location
Chr.I: 6436046 - 6441192 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_059452.8 NP_491853.2 6 - 7 958 154

View Full Product Details