Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
try-7
Entrez Gene ID
191201
Gene Name
Peptidase S1 domain-containing protein
Gene Aliases
CELE_ZC581.6
Chromosome Location
Chr.I: 6640651 - 6642643 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_059509.5 NP_491910.2 2 - 3 357 110

View Full Product Details