Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
lbp-6
Entrez Gene ID
172390
Gene Name
Fatty acid-binding protein homolog 6
Gene Aliases
CELE_W02D3.5
Chromosome Location
Chr.I: 6728290 - 6729376 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_059525.6 NP_491926.1 1 - 2 217 70

View Full Product Details