Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
kin-14
Entrez Gene ID
172440
Gene Name
Tyrosine-protein kinase
Gene Aliases
CELE_F22D6.1
Chromosome Location
Chr.I: 7075918 - 7079575 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_059603.8 NP_492004.1 8 - 9 1236 85

View Full Product Details