Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
xpa-1
Entrez Gene ID
187120
Gene Name
XPA_C domain-containing protein
Gene Aliases
CELE_K07G5.2
Chromosome Location
Chr.I: 7159714 - 7160917 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_059624.2 NP_492025.1 4 - 5 459 65

View Full Product Details