Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
gska-3
Entrez Gene ID
172683
Gene Name
Protein kinase domain-containing protein
Gene Aliases
CELE_C36B1.10
Chromosome Location
Chr.I: 8749616 - 8751563 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_059966.5 NP_492367.1 2 - 3 558 85

View Full Product Details