Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ced-12
Entrez Gene ID
172890
Gene Name
protein-coding
Gene Aliases
CELE_Y106G6E.5
Chromosome Location
Chr.I: 10222881 - 10227009 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001306347.3 NP_001293276.1 2 - 3 466 76
NM_060292.7 NP_492693.1 2 - 3 429 76
GenBank mRNA AF324505.1 - 2 - 3 450 76
AF416781.1 - 2 - 3 431 76

View Full Product Details