Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
nas-30
Entrez Gene ID
190803
Gene Name
protein-coding
Gene Aliases
CELE_Y95B8A.1
Chromosome Location
Chr.I: 938464 - 947710 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_058394.6 NP_490795.5 12 - 13 1820 60

View Full Product Details