Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
cutl-20
Entrez Gene ID
171678
Gene Name
protein-coding
Gene Aliases
CELE_F54A5.2
Chromosome Location
Chr.I: 963695 - 973133 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001383432.1 6 - 7 771 58
NM_058397.7 NP_490798.1 11 - 12 2039 58

View Full Product Details