Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
cogc-1
Entrez Gene ID
3565555
Gene Name
Conserved Oligomeric Golgi (COG) Component
Gene Aliases
CELE_Y54E10A.2
Chromosome Location
Chr.I: 3233391 - 3256159 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_058727.6 NP_491128.4 1 - 2 65 80

View Full Product Details