Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
hcp-4
Entrez Gene ID
171964
Gene Name
HoloCentric chromosome binding Protein
Gene Aliases
CELE_T03F1.9
Chromosome Location
Chr.I: 3862774 - 3866271 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_058843.8 NP_491244.1 3 - 4 315 67
GenBank mRNA AF321299.1 - 2 - 3 282 67

View Full Product Details