Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
fkb-5
Entrez Gene ID
171974
Gene Name
protein-coding
Gene Aliases
CELE_C50F2.6
Chromosome Location
Chr.I: 3898046 - 3899890 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001381792.1 NP_001367683.1 2 - 3 337 68
NM_001384047.1 2 - 3 301 68

View Full Product Details