Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
lfe-2
Entrez Gene ID
172128
Gene Name
protein-coding
Gene Aliases
CELE_C46H11.4
Chromosome Location
Chr.I: 5027155 - 5041163 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001025876.5 NP_001021047.1 3 - 4 1004 95
NM_001025877.6 NP_001021048.1 4 - 5 612 95
NM_001025878.7 NP_001021049.1 7 - 8 724 95
NM_001262849.3 NP_001249778.1 7 - 8 707 95
NM_001372686.1 NP_001359911.1 6 - 7 646 95

View Full Product Details