Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
lfe-2
Entrez Gene ID
172128
Gene Name
protein-coding
Gene Aliases
CELE_C46H11.4
Chromosome Location
Chr.I: 5027155 - 5041163 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001025876.5 NP_001021047.1 5 - 6 1629 103
NM_001025877.6 NP_001021048.1 6 - 7 1237 103
NM_001025878.7 NP_001021049.1 9 - 10 1349 103
NM_001262849.3 NP_001249778.1 9 - 10 1332 103
NM_001372686.1 NP_001359911.1 8 - 9 1271 103

View Full Product Details