Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
cey-3
Entrez Gene ID
172211
Gene Name
CSD_1 domain-containing protein
Gene Aliases
CELE_M01E11.5
Chromosome Location
Chr.I: 5571795 - 5572949 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001380298.2 NP_001367859.1 2 - 3 460 83

View Full Product Details