Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
exc-5
Entrez Gene ID
177692
Gene Name
protein-coding
Gene Aliases
CELE_C33D9.1
Chromosome Location
Chr.IV: 8799081 - 8813670 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001027868.5 NP_001023039.1 4 - 5 490 154
NM_001307099.3 NP_001294028.1 3 - 4 289 154
NM_001307100.4 NP_001294029.1 5 - 6 726 154
GenBank mRNA AB060647.1 - 4 - 5 543 154
AF339059.1 - 4 - 5 543 154

View Full Product Details