Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
paf-2
Entrez Gene ID
180704
Gene Name
protein-coding
Gene Aliases
CELE_C52B9.7
Chromosome Location
Chr.X: 4291475 - 4297741 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_171668.9 NP_741768.1 6 - 7 952 62
NM_171669.6 NP_741769.1 4 - 5 1329 62
GenBank mRNA AF386745.1 - 5 - 6 768 62

View Full Product Details