Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
nhx-5
Entrez Gene ID
181258
Gene Name
protein-coding
Gene Aliases
CELE_F57C7.2
Chromosome Location
Chr.X: 10576960 - 10581072 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001029549.4 NP_001024720.1 8 - 9 1077 81
NM_001373485.2 NP_001359642.1 8 - 9 1077 81
GenBank mRNA AF497828.1 - 8 - 9 1098 81
AF497829.1 - 8 - 9 1098 81

View Full Product Details