Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
pde-3
Entrez Gene ID
183981
Gene Name
protein-coding
Gene Aliases
CELE_E01F3.1
Chromosome Location
Chr.II: 14919705 - 14953353 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001267523.2 NP_001254452.1 4 - 5 829 107
NM_001267525.3 NP_001254454.1 3 - 4 507 107
NM_001306640.4 NP_001293569.1 5 - 6 824 107
NM_001306641.2 NP_001293570.1 4 - 5 829 107
NM_001306642.3 NP_001293571.1 7 - 8 1065 107
NM_001306643.3 NP_001293572.1 6 - 7 906 107
NM_001381668.1 NP_001367248.1 7 - 8 1065 107
NM_001381670.1 NP_001368261.1 5 - 6 765 107
NM_001383996.1 7 - 8 1129 107
NM_064683.6 NP_497084.2 2 - 3 615 107

View Full Product Details