Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
rrf-2
Entrez Gene ID
173092
Gene Name
RNA-dependent RNA polymerase Family
Gene Aliases
CELE_M01G12.12
Chromosome Location
Chr.I: 12098671 - 12106407 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_060656.4 NP_493057.1 10 - 11 2315 100

View Full Product Details