Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
oac-36
Entrez Gene ID
186422
Gene Name
Acyl_transf_3 domain-containing protein
Gene Aliases
CELE_F56H6.12
Chromosome Location
Chr.I: 12312325 - 12314962 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_060711.3 NP_493112.1 4 - 5 604 64

View Full Product Details