Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
sra-25
Entrez Gene ID
188926
Gene Name
Serpentine receptor class alpha-25
Gene Aliases
CELE_T26E3.9
Chromosome Location
Chr.I: 12686116 - 12688206 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_060810.2 NP_493211.2 3 - 4 658 127

View Full Product Details