Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
sra-24
Entrez Gene ID
188195
Gene Name
Serpentine receptor class alpha-24
Gene Aliases
CELE_T06G6.2
Chromosome Location
Chr.I: 12705097 - 12706983 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_060819.2 NP_493220.2 4 - 5 870 136

View Full Product Details