Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
unc-61
Entrez Gene ID
179976
Gene Name
protein-coding
Gene Aliases
CELE_Y50E8A.4
Chromosome Location
Chr.V: 14731064 - 14734237 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001047733.5 NP_001041198.2 4 - 5 472 64
NM_074237.8 NP_506638.2 4 - 5 470 64
NM_182356.5 NP_872156.2 5 - 6 649 64

View Full Product Details